missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,4-Dimethyl-3-oxovalerate 95.0+%, TCI America™

Catalog No. P103125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P103125ML 25 mL
P1031100ML 100 mL
P1031500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P103125ML Supplier TCI America Supplier No. P103125ML

Chemical Identifiers

CAS 55107-14-7
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00008847
InChI Key XTXCFTMJPRXBBC-UHFFFAOYSA-N
Synonym methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2
PubChem CID 99597
IUPAC Name methyl 4,4-dimethyl-3-oxopentanoate
SMILES CC(C)(C)C(=O)CC(=O)OC

Specifications

Color Yellow
Boiling Point 84°C
Quantity 25 mL
Formula Weight 158.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4,4-Dimethyl-3-oxovalerate
EINECSNumber (2)-1505
TSCA Yes