missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4,4-dimethyl-3-oxovalerate, 95%
CAS: 55107-14-7 | C8H14O3 | 158.197 g/mol
$95.33 - $361.89
Chemical Identifiers
| CAS | 55107-14-7 |
|---|---|
| Molecular Formula | C8H14O3 |
| Molecular Weight (g/mol) | 158.197 |
| MDL Number | MFCD00008847 |
| InChI Key | XTXCFTMJPRXBBC-UHFFFAOYSA-N |
| Synonym | methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2 |
| PubChem CID | 99597 |
| IUPAC Name | methyl 4,4-dimethyl-3-oxopentanoate |
| SMILES | CC(C)(C)C(=O)CC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0600706
|
Thermo Scientific Chemicals
L0600706 |
5 g |
N/A
|
|
|||||
|
AAL0600714
|
Thermo Scientific Chemicals
L0600714 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 55107-14-7 | |
| 158.197 | |
| XTXCFTMJPRXBBC-UHFFFAOYSA-N | |
| 99597 | |
| CC(C)(C)C(=O)CC(=O)OC |
| C8H14O3 | |
| MFCD00008847 | |
| methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2 | |
| methyl 4,4-dimethyl-3-oxopentanoate |
Specifications
| 55107-14-7 | |
| 67°C to 70°C (13 mmHg) | |
| C8H14O3 | |
| MFCD00008847 | |
| 1764796 | |
| XTXCFTMJPRXBBC-UHFFFAOYSA-N | |
| methyl 4,4-dimethyl-3-oxopentanoate | |
| 99597 | |
| 95% |
| 0.99 | |
| 77°C (170°F) | |
| 1.432 | |
| 5 g | |
| methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2 | |
| CC(C)(C)C(=O)CC(=O)OC | |
| 158.197 | |
| 158.2 | |
| Methyl 4,4-dimethyl-3-oxovalerate |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 259-481-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only