missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4,4,4-Trifluoroacetoacetate 96.0+%, TCI America™
$246.43 - $772.00
Chemical Identifiers
| CAS | 83643-84-9 |
|---|---|
| Molecular Formula | C5H5F3O3 |
| Molecular Weight (g/mol) | 170.087 |
| MDL Number | MFCD00041004 |
| InChI Key | LKMUBWWZTSZGGV-UHFFFAOYSA-N |
| Synonym | methyl trifluoroacetoacetate, methyl 4,4,4-trifluoroacetoacetate, 4,4,4-trifluoroacetoacetic acid methyl ester, methyl-4,4,4-trifluoro-3-oxobutanoate, butanoic acid, 4,4,4-trifluoro-3-oxo-, methyl ester, acmc-209psv, methyltri-fluoroacetoacetate, methyl4,4,4-trifluoroacetoacetate, methyl-4,4,4-trifluoroacetoacetate, methyl 3-oxo-4,4,4-trifluorobutyrate |
| PubChem CID | 550266 |
| IUPAC Name | methyl 4,4,4-trifluoro-3-oxobutanoate |
| SMILES | COC(=O)CC(=O)C(F)(F)F |
Chemical Identifiers
| 83643-84-9 | |
| 170.087 | |
| LKMUBWWZTSZGGV-UHFFFAOYSA-N | |
| 550266 | |
| COC(=O)CC(=O)C(F)(F)F |
| C5H5F3O3 | |
| MFCD00041004 | |
| methyl trifluoroacetoacetate, methyl 4,4,4-trifluoroacetoacetate, 4,4,4-trifluoroacetoacetic acid methyl ester, methyl-4,4,4-trifluoro-3-oxobutanoate, butanoic acid, 4,4,4-trifluoro-3-oxo-, methyl ester, acmc-209psv, methyltri-fluoroacetoacetate, methyl4,4,4-trifluoroacetoacetate, methyl-4,4,4-trifluoroacetoacetate, methyl 3-oxo-4,4,4-trifluorobutyrate | |
| methyl 4,4,4-trifluoro-3-oxobutanoate |
Specifications
| 83643-84-9 | |
| 116°C | |
| MFCD00041004 | |
| 3272 | |
| LKMUBWWZTSZGGV-UHFFFAOYSA-N | |
| methyl 4,4,4-trifluoro-3-oxobutanoate | |
| 550266 | |
| ≥96.0% (GC) | |
| Methyl 4,4,4-Trifluoroacetoacetate |
| Yellow | |
| C5H5F3O3 | |
| 5 g | |
| methyl trifluoroacetoacetate, methyl 4,4,4-trifluoroacetoacetate, 4,4,4-trifluoroacetoacetic acid methyl ester, methyl-4,4,4-trifluoro-3-oxobutanoate, butanoic acid, 4,4,4-trifluoro-3-oxo-, methyl ester, acmc-209psv, methyltri-fluoroacetoacetate, methyl4,4,4-trifluoroacetoacetate, methyl-4,4,4-trifluoroacetoacetate, methyl 3-oxo-4,4,4-trifluorobutyrate | |
| COC(=O)CC(=O)C(F)(F)F | |
| 170.087 | |
| 170.09 | |
| Liquid |
Safety and Handling
TSCA : No