missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-nitrobenzoate, 98%

CAS: 618-95-1 | C8H7NO4 | 181.15 g/mol

$134.84 - $134.84

Chemical Identifiers

CAS 618-95-1
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007250
InChI Key AXLYJLKKPUICKV-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
PubChem CID 69260
IUPAC Name methyl 3-nitrobenzoate
SMILES COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC168441000
SDS View Documents
Thermo Scientific Chemicals
168441000
100 g Plastic bottle
Each for $134.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

618-95-1
181.15
AXLYJLKKPUICKV-UHFFFAOYSA-N
69260
COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
C8H7NO4
MFCD00007250
3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
methyl 3-nitrobenzoate

Specifications

618-95-1
Beige
Authentic
Plastic bottle
O2NC6H4CO2CH3
100 g
10, 6435
Solubility in water: insoluble
COC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
181.15
181.15
Crystalline Powder
76°C to 80°C
279°C
97.5% min. (GC)
C8H7NO4
MFCD00007250
09, 378
3-nitrobenzoic acid methyl ester, methyl m-nitrobenzoate, methyl3-nitrobenzoate, benzoic acid, 3-nitro-, methyl ester, m-nitrobenzoic acid, methyl ester, benzoic acid, m-nitro-, methyl ester, 3-nitro-benzoic acid methyl ester, 3-nitro-benzoicacimethylester, acmc-209mxc, methyl 3-nitro-benzoate
AXLYJLKKPUICKV-UHFFFAOYSA-N
methyl 3-nitrobenzoate
69260
98%
Methyl 3-nitrobenzoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-573-

SDS
Documents

Documents

RUO – Research Use Only