Learn More
Methyl 3-methyl-2-furoate, 98%
CAS: 6141-57-7 | C7H8O3 | 140.14 g/mol
Supplier: Thermo Scientific Chemicals A1054109
Description
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.
Solubility
Insoluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents.
Specifications
| Methyl 3-methyl-2-furoate | |
| 33°C to 37°C | |
| 82°C (180°F) | |
| MFCD00014110 | |
| 116835 | |
| Insoluble in water. | |
| COC(=O)C1=C(C)C=CO1 | |
| 140.14 | |
| 140.14 |
| 6141-57-7 | |
| 60°C (1 mmHg) | |
| C7H8O3 | |
| 10 g | |
| methyl 3-methyl-2-furoate, methyl 3-methylfuroate, methyl 3-methyl-2-furancarboxylate, furancarboxylic acid, methyl-, methyl ester, 3-methyl-2-furancarboxylic acid methyl ester, 3-methyl-furan-2-carboxylic acid methyl ester, 2-furoic acid, 3-methyl-, methyl ester, acmc-20a9f6, methyl-3-methyl-2-furoate, 2-furancarboxylic acid, 3-methyl-, methyl ester | |
| AQQYRDKMXXSIMP-UHFFFAOYSA-N | |
| methyl 3-methylfuran-2-carboxylate | |
| 350231 | |
| 98% |
Chemical Identifiers
| 6141-57-7 | |
| 140.14 | |
| AQQYRDKMXXSIMP-UHFFFAOYSA-N | |
| 350231 | |
| COC(=O)C1=C(C)C=CO1 |
| C7H8O3 | |
| MFCD00014110 | |
| methyl 3-methyl-2-furoate, methyl 3-methylfuroate, methyl 3-methyl-2-furancarboxylate, furancarboxylic acid, methyl-, methyl ester, 3-methyl-2-furancarboxylic acid methyl ester, 3-methyl-furan-2-carboxylic acid methyl ester, 2-furoic acid, 3-methyl-, methyl ester, acmc-20a9f6, methyl-3-methyl-2-furoate, 2-furancarboxylic acid, 3-methyl-, methyl ester | |
| methyl 3-methylfuran-2-carboxylate |
Safety and Handling
EINECSNumber : 228-131-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only