missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-methyl-2-furoate, 98%
Description
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.
Solubility
Insoluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents.
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.
Solubility
Insoluble in water.
Notes
Store in a cool dry place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 33°C to 37°C |
| Boiling Point | 60°C (1 mmHg) |
| Flash Point | 82°C (180°F) |
| Quantity | 5 g |
| Beilstein | 116835 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 140.14 |
| Percent Purity | 98% |
| Chemical Name or Material | Methyl 3-methyl-2-furoate |
Safety and Handling
| EINECSNumber | 228-131-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only