accessibility menu, dialog, popup

UserName

CAS RN 6141-57-7

IUPAC Name: methyl 3-methylfuran-2-carboxylate
Synonyms: methyl 3-methylfuran-2-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: AQQYRDKMXXSIMP-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=CO1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)