missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-methoxybenzoate, 98%

Catalog No. AAL0904109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0904109 10 g
AAL0904118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0904109 Supplier Thermo Scientific Chemicals Supplier No. L0904109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5368-81-0 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5368-81-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017193
InChI Key DUKYPQBGYRJVAN-UHFFFAOYSA-N
Synonym methyl m-anisate, methyl m-methoxybenzoate, 3-methoxybenzoic acid methyl ester, m-anisic acid, methyl ester, benzoic acid, 3-methoxy-, methyl ester, 3-methoxybenoic acid methyl ester, m-methoxybenzoic acid methyl ester, mehtyl 3-methoxybenzoate, acmc-1asq5, m-anisic acid methyl ester
PubChem CID 79332
IUPAC Name methyl 3-methoxybenzoate
SMILES COC(=O)C1=CC=CC(OC)=C1

Specifications

Density 1.15
Boiling Point 238°C
Flash Point >110°C (230°F)
Refractive Index 1.527
Quantity 10 g
Beilstein 2208567
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material Methyl 3-methoxybenzoate
EINECSNumber 226-359-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only