missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Methoxy-2-nitrobenzoate 98.0+%, TCI America™

Catalog No. M13641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13641G 1 g
M13645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13641G Supplier TCI America Supplier No. M13641G

Chemical Identifiers

CAS 5307-17-5
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD00051968
InChI Key FDQQRLPHAAICCR-UHFFFAOYSA-N
Synonym benzoic acid, 3-methoxy-2-nitro-, methyl ester, 3-methoxy-2-nitrobenzoic acid methyl ester, m-anisic acid, 2-nitro-, methyl ester, maybridge1_002124, cbmicro_008502, methyl 2-nitro-m-anisate, methyl 3-methoxy-2-nitro-benzoate, 2-nitro-m-anisic acid methyl ester, 3-methoxy-2-nitro-benzoic acid methyl ester
PubChem CID 79195
IUPAC Name methyl 3-methoxy-2-nitrobenzoate
SMILES COC(=O)C1=CC=CC(OC)=C1[N+]([O-])=O

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 1 g
Formula Weight 211.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Methoxy-2-nitrobenzoate