missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-cyclohexenecarboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 452861000
| Quantity | 100mL |
|---|
Chemical Identifiers
| C8H12O2 | |
| MFCD00053266 | |
| methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity | |
| methyl cyclohex-3-ene-1-carboxylate |
Specifications
| Methyl 3-cyclohexenecarboxylate | |
| 1.0200g/mL | |
| 60°C | |
| 96% min. (GC) | |
| MFCD00053266 | |
| 1.02 | |
| IPUNVLFESXFVFH-UHFFFAOYNA-N | |
| methyl cyclohex-3-ene-1-carboxylate | |
| 96926 | |
| 97% |
| 6493-77-2 | |
| 185.0°C | |
| Authentic | |
| C8H12O2 | |
| 100mL | |
| methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity | |
| COC(=O)C1CCC=CC1 | |
| 140.18 | |
| 140.18 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 229-376-6
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.