accessibility menu, dialog, popup

UserName

CAS RN 6493-77-2

IUPAC Name: methyl cyclohex-3-ene-1-carboxylate
Synonyms: methyl cyclohex-3-ene-1-carboxylate
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: IPUNVLFESXFVFH-UHFFFAOYNA-N
SMILES: COC(=O)C1CCC=CC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)