missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyclohexenecarboxylate, 97%, Thermo Scientific™

Catalog No. p-7077397
Change view
Click to view available options
Quantity:
25mL
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC452860250 25mL
AC452861000 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC452860250 Supplier Thermo Scientific Chemicals Supplier No. 452860250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6493-77-2
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00053266
InChI Key IPUNVLFESXFVFH-UHFFFAOYNA-N
Synonym methyl cyclohex-3-enecarboxylate, 3-cyclohexene-1-carboxylic acid methyl ester, methyl 3-cyclohexenecarboxylate, 3-cyclohexene-1-carboxylic acid, methyl ester, methyl 3-cyclohexene-1-carboxylate, 1-carbomethoxy-3-cyclohexene, methyl 4-cyclohexenecarboxylate, methyl 1,2,3,6-tetrahydrobenzoate, 1,3-butadiene-methyl acrylate adduct, acmc-1bity
PubChem CID 96926
IUPAC Name methyl cyclohex-3-ene-1-carboxylate
SMILES COC(=O)C1CCC=CC1

Specifications

Density 1.0200g/mL
Boiling Point 185.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25mL
Specific Gravity 1.02
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material Methyl 3-cyclohexenecarboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 229-376-6

RUO – Research Use Only