missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromophenylacetate 98.0+%, TCI America™

Catalog No. p-7133443
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28045G 5 g
M280425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28045G Supplier TCI America Supplier No. M28045G

Chemical Identifiers

CAS 150529-73-0
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD06858764
InChI Key ULSSGHADTSRELG-UHFFFAOYSA-N
Synonym 3-Bromophenylacetic Acid Methyl Ester
PubChem CID 11746402
IUPAC Name methyl 2-(3-bromophenyl)acetate
SMILES COC(=O)CC1=CC(=CC=C1)Br

Specifications

Color Yellow
Boiling Point 268°C
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Bromophenylacetate