accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 150529-73-0

IUPAC Name: methyl 2-(3-bromophenyl)acetate
Synonyms: methyl 2-(3-bromophenyl)acetate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: ULSSGHADTSRELG-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC(Br)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results