missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-5-hydroxybenzoate, 97%, Thermo Scientific Chemicals

Catalog No. AAH5172403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5172403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5172403 Supplier Thermo Scientific Chemicals Supplier No. H5172403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 192810-12-1 | Molecular Formula: C8H7BrO3 | Molecular Weight: 231.045 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 192810-12-1
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD10566800
InChI Key KRTBWIIGEJIUSA-UHFFFAOYSA-N
Synonym 3-bromo-5-hydroxybenzoic acid methyl ester, acmc-209f1w, methyl3-bromo-5-hydroxybenzoate, methyl 3-bromo-5-hydroxylbenzoate, 3-bromo-5-hydroxy-benzoic acid methyl ester, benzoic acid,3-bromo-5-hydroxy-,methyl ester, benzoic acid, 3-bromo-5-hydroxy-, methyl ester
PubChem CID 53216607
IUPAC Name methyl 3-bromo-5-hydroxybenzoate
SMILES COC(=O)C1=CC(=CC(=C1)Br)O

Specifications

Melting Point 130°C to 135°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 231.05
Percent Purity 97%
Chemical Name or Material Methyl 3-bromo-5-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only