CAS RN 192810-12-1
IUPAC Name:
methyl 3-bromo-5-hydroxybenzoate
Synonyms:
methyl 3-bromo-5-hydroxybenzoate
Molecular Weight (g/mol):
231.05
Molecular Formula:
C8H7BrO3
InChi Key:
KRTBWIIGEJIUSA-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(O)=CC(Br)=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 3-bromo-5-hydroxybenzoate, 97%, Thermo Scientific Chemicals
Methyl 3-bromo-5-hydroxybenzoate, 97%, Thermo Scientific Chemicals
CAS: 192810-12-1 Molecular Formula: C8H7BrO3 Molecular Weight (g/mol): 231.045 MDL Number: MFCD10566800 InChI Key: KRTBWIIGEJIUSA-UHFFFAOYSA-N Synonym: 3-bromo-5-hydroxybenzoic acid methyl ester,acmc-209f1w,methyl3-bromo-5-hydroxybenzoate,methyl 3-bromo-5-hydroxylbenzoate,3-bromo-5-hydroxy-benzoic acid methyl ester,benzoic acid,3-bromo-5-hydroxy-,methyl ester,benzoic acid, 3-bromo-5-hydroxy-, methyl ester PubChem CID: 53216607 IUPAC Name: methyl 3-bromo-5-hydroxybenzoate SMILES: COC(=O)C1=CC(=CC(=C1)Br)O