missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-5-fluorophenylacetate, 96%

Catalog No. AAH6256106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6256106 5 g
AAH6256103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6256106 Supplier Thermo Scientific Chemicals Supplier No. H6256106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 937732-38-2 | C9H8BrFO2 | 247.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 937732-38-2
Molecular Formula C9H8BrFO2
Molecular Weight (g/mol) 247.063
InChI Key AJSCFDHWZUZCNE-UHFFFAOYSA-N
Synonym methyl 2-3-bromo-5-fluorophenyl acetate, methyl 3-bromo-5-fluorophenyl acetate
PubChem CID 59290393
IUPAC Name methyl 2-(3-bromo-5-fluorophenyl)acetate
SMILES COC(=O)CC1=CC(=CC(=C1)Br)F

Specifications

Quantity 5 g
Formula Weight 247.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 3-bromo-5-fluorophenylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only