accessibility menu, dialog, popup

UserName

CAS RN 937732-38-2

IUPAC Name: methyl 2-(3-bromo-5-fluorophenyl)acetate
Synonyms: methyl 2-(3-bromo-5-fluorophenyl)acetate
Molecular Weight (g/mol): 247.06
Molecular Formula: C9H8BrFO2
InChi Key: AJSCFDHWZUZCNE-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC(F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results