missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-5-fluorobenzoate, 98%

Catalog No. p-7031253
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3226203 1 g
AAH3226206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3226203 Supplier Thermo Scientific Chemicals Supplier No. H3226203
Only null left

CAS: 334792-52-8 | C8H6BrFO2 | 233.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 334792-52-8
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.04
MDL Number MFCD07780735
InChI Key JERAACCIOWRRQA-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-5-fluoro-, methyl ester, acmc-209i1c, methyl 5-bromo-3-fluorobenzoate, methyl-3-bromo-5-fluorobenzoate, 3-bromo-5-fluoro-benzoic acid methyl ester, benzoicacid, 3-bromo-5-fluoro-, methyl ester
PubChem CID 21942598
IUPAC Name methyl 3-bromo-5-fluorobenzoate
SMILES COC(=O)C1=CC(F)=CC(Br)=C1

Specifications

Refractive Index 1.5345
Quantity 1 g
Formula Weight 233.04
Percent Purity 98%
Chemical Name or Material Methyl 3-bromo-5-fluorobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only