CAS RN 334792-52-8
IUPAC Name:
methyl 3-bromo-5-fluorobenzoate
Synonyms:
methyl 3-bromo-5-fluorobenzoate
Molecular Weight (g/mol):
233.04
Molecular Formula:
C8H6BrFO2
InChi Key:
JERAACCIOWRRQA-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(F)=CC(Br)=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 3-bromo-5-fluorobenzoate, 98%
Methyl 3-bromo-5-fluorobenzoate, 98%
CAS: 334792-52-8 Molecular Formula: C8H6BrFO2 Molecular Weight (g/mol): 233.04 MDL Number: MFCD07780735 InChI Key: JERAACCIOWRRQA-UHFFFAOYSA-N Synonym: 3-bromo-5-fluorobenzoic acid methyl ester,benzoic acid, 3-bromo-5-fluoro-, methyl ester,acmc-209i1c,methyl 5-bromo-3-fluorobenzoate,methyl-3-bromo-5-fluorobenzoate,3-bromo-5-fluoro-benzoic acid methyl ester,benzoicacid, 3-bromo-5-fluoro-, methyl ester PubChem CID: 21942598 IUPAC Name: methyl 3-bromo-5-fluorobenzoate SMILES: COC(=O)C1=CC(F)=CC(Br)=C1