missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-4-methylbenzoate 98.0+%, TCI America™

Catalog No. m218125g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M218125G 25 g
M21815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M218125G Supplier TCI America Supplier No. M218125G
Only null left

Chemical Identifiers

CAS 104901-43-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00144769
InChI Key MASRAGFWFYHMFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoic acid methyl ester, methyl 3-bromo-p-toluate, methyl3-bromo-4-methylbenzoate, benzoic acid, 3-bromo-4-methyl-, methyl ester, 3-bromo-p-toluic acid methyl ester, 3-bromo-4-methyl methyl benzoate, 3-bromo-4-methyl-benzoic acid methyl ester, pubchem3967, acmc-2098dw, methyl 3-bromo4-methylbenzoate
PubChem CID 7015747
IUPAC Name methyl 3-bromo-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)Br

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 229.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 3-Bromo-4-methylbenzoate