accessibility menu, dialog, popup

UserName

CH3OOCH3Br

CAS RN 104901-43-1

IUPAC Name: methyl 3-bromo-4-methylbenzoate
Synonyms: methyl 3-bromo-4-methylbenzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: MASRAGFWFYHMFI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C(Br)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)