missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-bromo-4-methylbenzoate, 99%
Description
It is a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a pharmaceutical intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at room temperature. Incompatible with oxidizing agents.
It is a pharmaceutical intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 5 g |
| Solubility Information | Soluble in dimethyl sulfoxide. |
| Formula Weight | 229.07 |
| Percent Purity | 99% |
| Chemical Name or Material | Methyl 3-bromo-4-methylbenzoate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only