missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-4-methylbenzoate, 99%

Catalog No. AAB2437206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2437206 5 g
AAB2437214 25 g
AAB2437222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2437206 Supplier Thermo Scientific Chemicals Supplier No. B2437206

CAS: 104901-43-1 | C9H9BrO2 | 229.073 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 104901-43-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00144769
InChI Key MASRAGFWFYHMFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoic acid methyl ester, methyl 3-bromo-p-toluate, methyl3-bromo-4-methylbenzoate, benzoic acid, 3-bromo-4-methyl-, methyl ester, 3-bromo-p-toluic acid methyl ester, 3-bromo-4-methyl methyl benzoate, 3-bromo-4-methyl-benzoic acid methyl ester, pubchem3967, acmc-2098dw, methyl 3-bromo4-methylbenzoate
PubChem CID 7015747
IUPAC Name methyl 3-bromo-4-methylbenzoate
SMILES CC1=C(C=C(C=C1)C(=O)OC)Br

Specifications

Quantity 5 g
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 229.07
Percent Purity 99%
Chemical Name or Material Methyl 3-bromo-4-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only