missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-4-hydroxybenzoate, 98%

Catalog No. p-7030545
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3022209 10 g
AAH3022218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3022209 Supplier Thermo Scientific Chemicals Supplier No. H3022209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29415-97-2 | C8H7BrO3 | 231.05 g/mol

A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 29415-97-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD06203850
InChI Key RKUNSPWAQIUGEZ-UHFFFAOYSA-N
Synonym methyl3-bromo-4-hydroxybenzoate, 3-bromo-4-hydroxybenzoic acid methyl ester, benzoic acid, 3-bromo-4-hydroxy-, methyl ester, 2-bromo-4-methoxycarbonyl phenol, methyl 4-hydroxy-3-bromobenzoate, methyl-3-bromo-4-hydroxybenzoate, methyl 3-bromo-4-hydroxy-benzoate, methyl 3-bromo-4-hydroxybenzoate, methyl 3-bromanyl-4-oxidanyl-benzoate, 3-bromo-4-hydroxy-benzoic acid methyl ester
PubChem CID 4778958
IUPAC Name methyl 3-bromo-4-hydroxybenzoate
SMILES COC(=O)C1=CC(Br)=C(O)C=C1

Specifications

Melting Point 108°C to 110°C
Quantity 10 g
Formula Weight 231.05
Percent Purity 98%
Chemical Name or Material Methyl 3-bromo-4-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only