accessibility menu, dialog, popup

UserName

CAS RN 29415-97-2

IUPAC Name: methyl 3-bromo-4-hydroxybenzoate
Synonyms: methyl 3-bromo-4-hydroxybenzoate
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: RKUNSPWAQIUGEZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results