missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-4-fluorobenzoate 98.0+%, TCI America™

Catalog No. p-7133536
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M30181G 1 g
M30185G 5 g
2 options

Catalog No. M30181G

Supplier: TCI America M30181G

Only null left

Chemical Identifiers

CAS 82702-31-6
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.04
MDL Number MFCD04972915
InChI Key JVORYGNKYAXATM-UHFFFAOYSA-N
Synonym 3-Bromo-4-fluorobenzoic Acid Methyl Ester
PubChem CID 7018022
IUPAC Name methyl 3-bromo-4-fluorobenzoate
SMILES COC(=O)C1=CC=C(F)C(Br)=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 233.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 3-Bromo-4-fluorobenzoate