missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-amino-5-fluorobenzoate, 98%
CAS: 884497-46-5 | C8H8FNO2 | 169.155 g/mol
$57.73 - $677.88
Chemical Identifiers
| CAS | 884497-46-5 |
|---|---|
| Molecular Formula | C8H8FNO2 |
| Molecular Weight (g/mol) | 169.155 |
| MDL Number | MFCD05863683 |
| InChI Key | YKEJVXMOMKWEPV-UHFFFAOYSA-N |
| Synonym | methyl3-amino-5-fluorobenzoate, methyl 5-amino-3-fluorobenzoate, 5-amino-3-fluorobenzoic acid methyl ester, 3-amino-5-fluoro-benzoic acid methyl ester |
| PubChem CID | 3807862 |
| IUPAC Name | methyl 3-amino-5-fluorobenzoate |
| SMILES | COC(=O)C1=CC(=CC(=C1)F)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH63038MD
|
Thermo Scientific Chemicals
H63038MD |
250 mg |
Each for $57.73
|
|
|||||
|
AAH6303803
|
Thermo Scientific Chemicals
H6303803 |
1 g |
Each for $159.40
|
|
|||||
|
AAH6303806
|
Thermo Scientific Chemicals
H6303806 |
5 g |
Each for $677.88
|
|
|||||
Description
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 884497-46-5 | |
| 169.155 | |
| YKEJVXMOMKWEPV-UHFFFAOYSA-N | |
| 3807862 | |
| COC(=O)C1=CC(=CC(=C1)F)N |
| C8H8FNO2 | |
| MFCD05863683 | |
| methyl3-amino-5-fluorobenzoate, methyl 5-amino-3-fluorobenzoate, 5-amino-3-fluorobenzoic acid methyl ester, 3-amino-5-fluoro-benzoic acid methyl ester | |
| methyl 3-amino-5-fluorobenzoate |
Specifications
| 884497-46-5 | |
| C8H8FNO2 | |
| 250 mg | |
| Slightly soluble in water. | |
| COC(=O)C1=CC(=CC(=C1)F)N | |
| 169.155 | |
| 169.16 | |
| Solid |
| White to Yellow | |
| MFCD05863683 | |
| methyl3-amino-5-fluorobenzoate, methyl 5-amino-3-fluorobenzoate, 5-amino-3-fluorobenzoic acid methyl ester, 3-amino-5-fluoro-benzoic acid methyl ester | |
| YKEJVXMOMKWEPV-UHFFFAOYSA-N | |
| methyl 3-amino-5-fluorobenzoate | |
| 3807862 | |
| 98% | |
| Methyl 3-amino-5-fluorobenzoate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only