missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Amino-2-fluorobenzoate 96.0+%, TCI America™

Catalog No. m28681g
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28681G 1 g
M2868200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28681G Supplier TCI America Supplier No. M28681G
Only null left

Chemical Identifiers

CAS 1195768-18-3
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD12923265
InChI Key UOYDNSRSUSNCKS-UHFFFAOYSA-N
Synonym 3-Amino-2-fluorobenzoic Acid Methyl Ester
PubChem CID 46311080
IUPAC Name methyl 3-amino-2-fluorobenzoate
SMILES COC(=O)C1=C(C(=CC=C1)N)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 169.16
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl 3-Amino-2-fluorobenzoate