accessibility menu, dialog, popup

UserName

CH3OONH2F

CAS RN 1195768-18-3

IUPAC Name: methyl 3-amino-2-fluorobenzoate
Synonyms: methyl 3-amino-2-fluorobenzoate
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: UOYDNSRSUSNCKS-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(F)C(N)=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results