missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromobenzoate, 98%, Thermo Scientific Chemicals

Catalog No. p-7025273
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2066806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2066806 Supplier Thermo Scientific Chemicals Supplier No. B2066806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51329-15-8 | Molecular Formula: C8H6Br2O2 | Molecular Weight: 293.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51329-15-8
Molecular Formula C8H6Br2O2
Molecular Weight (g/mol) 293.94
MDL Number MFCD00082648
InChI Key GSMAWUZTAIOCPL-UHFFFAOYSA-N
Synonym methyl3,5-dibromobenzoate, 3,5-dibromobenzoic acid methyl ester, benzoic acid, 3,5-dibromo-, methyl ester, 3,5-dibromomethylbenzoate, 1,3-dibromo-5-methoxycarbonyl benzene, pubchem3973, maybridge1_007706, acmc-209kt2, ksc274i7j, rarechem al bf 0770
PubChem CID 621923
IUPAC Name methyl 3,5-dibromobenzoate
SMILES COC(=O)C1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 61°C to 64°C
Quantity 5 g
Beilstein 2255855
Formula Weight 293.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromobenzoate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only