accessibility menu, dialog, popup

UserName

CAS RN 51329-15-8

IUPAC Name: methyl 3,5-dibromobenzoate
Synonyms: methyl 3,5-dibromobenzoate
Molecular Weight (g/mol): 293.94
Molecular Formula: C8H6Br2O2
InChi Key: GSMAWUZTAIOCPL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC(Br)=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)