missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-Dibromo-4-methylbenzoate 98.0+%, TCI America™

Catalog No. p-7127547
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28715G 5 g
D287125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28715G Supplier TCI America Supplier No. D28715G

Chemical Identifiers

CAS 74896-66-5
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.97
MDL Number MFCD00010729
InChI Key SRLAXDDPVYZYNI-UHFFFAOYSA-N
Synonym 3,5-dibromo-4-methylbenzoic acid methyl ester, methyl 3,5-dibromo-p-toluate, 3,5-dibromo-p-toluic acid methyl ester, pubchem4611, methyl3,5-dibromo-4-methylbenzoate, acmc-1bm25, zerenex e/9072914, methyl 3,5-dibromo-4-methyl-benzoate, methyl 3,5-bis bromanyl-4-methyl-benzoate, 2,6-dibromo-4-methoxycarbonyl toluene
PubChem CID 624016
IUPAC Name methyl 3,5-dibromo-4-methylbenzoate
SMILES COC(=O)C1=CC(Br)=C(C)C(Br)=C1

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 5 g
Formula Weight 307.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Dibromo-4-methylbenzoate