accessibility menu, dialog, popup

UserName

CAS RN 74896-66-5

IUPAC Name: methyl 3,5-dibromo-4-methylbenzoate
Synonyms: methyl 3,5-dibromo-4-methylbenzoate
Molecular Weight (g/mol): 307.97
Molecular Formula: C9H8Br2O2
InChi Key: SRLAXDDPVYZYNI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=C(C)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results