missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-di-tert-butyl-4-hydroxybenzoate, 98%

Catalog No. AAA1483714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1483714 25 g
AAA1483722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1483714 Supplier Thermo Scientific Chemicals Supplier No. A1483714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2511-22-0 | C16H24O3 | 264.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2511-22-0
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.37
MDL Number MFCD00017253
InChI Key UPVYFJALDJUSOV-UHFFFAOYSA-N
Synonym methyl 3,5-di-tert-butyl-4-hydroxybenzoate, benzoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy-, methyl ester, 3,5-di-tert-butyl-4-hydroxybenzoic acid methyl ester, 3,5-bis 1,1-dimethylethyl-4-hydroxy-benzoic acid ethyl ester, acmc-20akhp, methyl 3,5-bis-tert-butyl-4-hydroxybenzoate, benzoic acid, 3,5-di-tert-butyl-4-hydroxy-, methyl ester, methyl 3,5-di-tert.butyl-4-hydroxylbenzoate, methyl 3,5-bis tert-butyl-4-hydroxybenzoate
PubChem CID 75639
IUPAC Name methyl 3,5-ditert-butyl-4-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 164°C to 166°C
Quantity 25 g
Beilstein 2218125
Formula Weight 264.37
Percent Purity 98%
Chemical Name or Material Methyl 3,5-di-tert-butyl-4-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-729-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only