missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-Dihydroxy-2-methylbenzoate 98.0+%, TCI America™

Catalog No. M27131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27131G Supplier TCI America Supplier No. M27131G
Only null left

Chemical Identifiers

CAS 740799-82-0
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD16250128
InChI Key ONNFZKHTMFVKNY-UHFFFAOYSA-N
Synonym 3,4-Dihydroxy-2-methylbenzoic Acid Methyl Ester
PubChem CID 59016598
IUPAC Name methyl 3,4-dihydroxy-2-methylbenzoate
SMILES COC(=O)C1=C(C)C(O)=C(O)C=C1

Specifications

Melting Point 161°C
Quantity 1 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4-Dihydroxy-2-methylbenzoate