accessibility menu, dialog, popup

UserName

CH3HOHOOOCH3

CAS RN 740799-82-0

IUPAC Name: methyl 3,4-dihydroxy-2-methylbenzoate
Synonyms: methyl 3,4-dihydroxy-2-methylbenzoate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: ONNFZKHTMFVKNY-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C(O)=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results