missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-bromophenyl)propionate, 97%

Catalog No. AAH6154606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6154606 5 g
AAH6154614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6154606 Supplier Thermo Scientific Chemicals Supplier No. H6154606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75567-84-9 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75567-84-9
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD09953150
InChI Key FKPYNBFWCSTPOT-UHFFFAOYSA-N
Synonym methyl 3-4-bromophenyl propanoate, methyl 3-4-bromophenyl propionate, 3-4-bromophenyl propanoic acid methyl ester, benzenepropanoic acid, 4-bromo-, methyl ester, 3-4-bromophenyl propionic acid methyl ester, acmc-209oyw, 3-4-bromophenyl propionate, ksc498a6l, methyl3-4-bromophenyl propionate, 3-4-bromophenyl-propionic acid methyl ester
PubChem CID 11447883
IUPAC Name methyl 3-(4-bromophenyl)propanoate
SMILES COC(=O)CCC1=CC=C(C=C1)Br

Specifications

Color Colorless to Yellow
Quantity 5 g
Formula Weight 243.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-(4-bromophenyl)propionate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only