accessibility menu, dialog, popup

UserName

CAS RN 75567-84-9

H3COOBr

CAS RN 75567-84-9

IUPAC Name: methyl 3-(4-bromophenyl)propanoate
Synonyms: methyl 3-(4-bromophenyl)propanoate
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: FKPYNBFWCSTPOT-UHFFFAOYSA-N
SMILES: COC(=O)CCC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)