missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(Trifluoromethyl)benzoate 98.0+%, TCI America™

Catalog No. M24755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24755G 5 g
M247525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24755G Supplier TCI America Supplier No. M24755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 344-96-7
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00236050
InChI Key JLCAMDSWNZBTEE-UHFFFAOYSA-N
Synonym methyl 2-trifluoromethyl benzoate, benzoic acid, trifluoromethyl-, methyl ester, acmc-1ahfp, methyl o-trifluoromethylbenzoate, jlcamdswnzbtee-uhfffaoysa, methyl-2-trifluoromethyl benzoate, 2-trifluoromethylbenzoic acid methyl ester, 2-trifluoromethyl-benzoic acid methyl ester, 2-trifluoromethyl benzoic acid methyl ester, methyl alpha,alpha,alpha-trifluoro-o-toluate
PubChem CID 2775578
IUPAC Name methyl 2-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

Color Colorless
Boiling Point 105°C
Quantity 5 g
Formula Weight 204.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-(Trifluoromethyl)benzoate