accessibility menu, dialog, popup

UserName

CH3OOFFF

CAS RN 344-96-7

IUPAC Name: methyl 2-(trifluoromethyl)benzoate
Synonyms: methyl 2-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: JLCAMDSWNZBTEE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results