missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7133437
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27975G 5 g
M279725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27975G Supplier TCI America Supplier No. M27975G

Chemical Identifiers

CAS 57113-90-3
Molecular Formula C13H16N2O6
Molecular Weight (g/mol) 296.279
MDL Number MFCD06797690
InChI Key VEDIIGMQOAWKGH-UHFFFAOYSA-N
Synonym 2-(tert-Butoxycarbonylamino)-3-nitrobenzoic Acid Methyl Ester, Methyl 2-(Boc-amino)-3-nitrobenzoate, 2-(Boc-amino)-3-nitrobenzoic Acid Methyl Ester
PubChem CID 9965968
IUPAC Name methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-nitrobenzoate
SMILES CC(C)(C)OC(=O)NC1=C(C=CC=C1[N+](=O)[O-])C(=O)OC

Specifications

Melting Point 106°C
Color Yellow
Quantity 5 g
Formula Weight 296.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate