accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHNOOOOCH3

CAS RN 57113-90-3

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}-3-nitrobenzoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]-3-nitrobenzoate
Molecular Weight (g/mol): 296.28
Molecular Formula: C13H16N2O6
InChi Key: VEDIIGMQOAWKGH-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(NC(=O)OC(C)(C)C)C(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results