missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Pyridylacetate 97.0+%, TCI America™
Supplier: TCI America P12685G
Specifications
| Methyl 2-Pyridylacetate | |
| Yellow | |
| C8H9NO2 | |
| 5 g | |
| ORAKNQSHWMHCEY-UHFFFAOYSA-N | |
| methyl 2-(pyridin-2-yl)acetate | |
| 74264 | |
| ≥97.0% (GC) |
| 1658-42-0 | |
| 137°C | |
| MFCD00006358 | |
| methyl 2-pyridylacetate, methyl 2-pyridin-2-yl acetate, 2-pyridylacetic acid methyl ester, 2-pyridineacetic acid, methyl ester, methyl 2-pyridinylacetate, methyl pyridin-2-ylacetate, methyl 2-pyridineacetate, 2-pyridineacetic acid methyl ester, pyridin-2-yl-acetic acid methyl ester, methyl pyridine-2-acetate | |
| COC(=O)CC1=CC=CC=N1 | |
| 151.17 | |
| 151.17 | |
| Liquid |
Chemical Identifiers
| 1658-42-0 | |
| 151.17 | |
| ORAKNQSHWMHCEY-UHFFFAOYSA-N | |
| 74264 | |
| COC(=O)CC1=CC=CC=N1 |
| C8H9NO2 | |
| MFCD00006358 | |
| methyl 2-pyridylacetate, methyl 2-pyridin-2-yl acetate, 2-pyridylacetic acid methyl ester, 2-pyridineacetic acid, methyl ester, methyl 2-pyridinylacetate, methyl pyridin-2-ylacetate, methyl 2-pyridineacetate, 2-pyridineacetic acid methyl ester, pyridin-2-yl-acetic acid methyl ester, methyl pyridine-2-acetate | |
| methyl 2-(pyridin-2-yl)acetate |
Safety and Handling
TSCA : No