missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Pyridylacetate 97.0+%, TCI America™
$126.76 - $380.27
Chemical Identifiers
| CAS | 1658-42-0 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.17 |
| MDL Number | MFCD00006358 |
| InChI Key | ORAKNQSHWMHCEY-UHFFFAOYSA-N |
| Synonym | methyl 2-pyridylacetate, methyl 2-pyridin-2-yl acetate, 2-pyridylacetic acid methyl ester, 2-pyridineacetic acid, methyl ester, methyl 2-pyridinylacetate, methyl pyridin-2-ylacetate, methyl 2-pyridineacetate, 2-pyridineacetic acid methyl ester, pyridin-2-yl-acetic acid methyl ester, methyl pyridine-2-acetate |
| PubChem CID | 74264 |
| IUPAC Name | methyl 2-(pyridin-2-yl)acetate |
| SMILES | COC(=O)CC1=CC=CC=N1 |
Chemical Identifiers
| 1658-42-0 | |
| 151.17 | |
| ORAKNQSHWMHCEY-UHFFFAOYSA-N | |
| 74264 | |
| COC(=O)CC1=CC=CC=N1 |
| C8H9NO2 | |
| MFCD00006358 | |
| methyl 2-pyridylacetate, methyl 2-pyridin-2-yl acetate, 2-pyridylacetic acid methyl ester, 2-pyridineacetic acid, methyl ester, methyl 2-pyridinylacetate, methyl pyridin-2-ylacetate, methyl 2-pyridineacetate, 2-pyridineacetic acid methyl ester, pyridin-2-yl-acetic acid methyl ester, methyl pyridine-2-acetate | |
| methyl 2-(pyridin-2-yl)acetate |
Specifications
| 1658-42-0 | |
| 137°C | |
| MFCD00006358 | |
| methyl 2-pyridylacetate, methyl 2-pyridin-2-yl acetate, 2-pyridylacetic acid methyl ester, 2-pyridineacetic acid, methyl ester, methyl 2-pyridinylacetate, methyl pyridin-2-ylacetate, methyl 2-pyridineacetate, 2-pyridineacetic acid methyl ester, pyridin-2-yl-acetic acid methyl ester, methyl pyridine-2-acetate | |
| COC(=O)CC1=CC=CC=N1 | |
| 151.17 | |
| 151.17 | |
| Liquid |
| Yellow | |
| C8H9NO2 | |
| 5 g | |
| ORAKNQSHWMHCEY-UHFFFAOYSA-N | |
| methyl 2-(pyridin-2-yl)acetate | |
| 74264 | |
| ≥97.0% (GC) | |
| Methyl 2-Pyridylacetate |
Safety and Handling
TSCA : No