missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%

Catalog No. p-7044396
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0618809 10 g
AAL0618818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0618809 Supplier Thermo Scientific Chemicals Supplier No. L0618809

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5081-42-5 | C11H13NO7 | 271.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5081-42-5
Molecular Formula C11H13NO7
Molecular Weight (g/mol) 271.225
MDL Number MFCD00007097
InChI Key HCEIEWYPDCDNNU-UHFFFAOYSA-N
Synonym methyl 2-nitro-3,4,5-trimethoxybenzoate, methyl 6-nitro-3,4,5-trimethoxybenzoate, 3,4,5-trimethoxy-2-nitrobenzoic acid methyl ester, benzoic acid, 3,4,5-trimethoxy-2-nitro-, methyl ester, 2-nitro-3,4,5-trimethoxybenzoic acid methyl ester, pubchem22545, opera_id_1985, acmc-1ar8w, hceiewypdcdnnu-uhfffaoysa, methyl 3,5-trimethoxy-2-nitrobenzoate
PubChem CID 78765
IUPAC Name methyl 3,4,5-trimethoxy-2-nitrobenzoate
SMILES COC1=C(C(=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC)OC

Specifications

Melting Point 66°C to 70°C
Quantity 10 g
Beilstein 2702653
Formula Weight 271.23
Percent Purity 98%
Chemical Name or Material Methyl 2-nitro-3,4,5-trimethoxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-794-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only