CAS RN 5081-42-5
IUPAC Name:
methyl 3,4,5-trimethoxy-2-nitrobenzoate
Synonyms:
methyl 3,4,5-trimethoxy-2-nitrobenzoate
Molecular Weight (g/mol):
271.23
Molecular Formula:
C11H13NO7
InChi Key:
HCEIEWYPDCDNNU-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(OC)=C(OC)C(OC)=C1[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%
Methyl 2-nitro-3,4,5-trimethoxybenzoate, 98%
CAS: 5081-42-5 Molecular Formula: C11H13NO7 Molecular Weight (g/mol): 271.225 MDL Number: MFCD00007097 InChI Key: HCEIEWYPDCDNNU-UHFFFAOYSA-N Synonym: methyl 2-nitro-3,4,5-trimethoxybenzoate,methyl 6-nitro-3,4,5-trimethoxybenzoate,3,4,5-trimethoxy-2-nitrobenzoic acid methyl ester,benzoic acid, 3,4,5-trimethoxy-2-nitro-, methyl ester,2-nitro-3,4,5-trimethoxybenzoic acid methyl ester,pubchem22545,opera_id_1985,acmc-1ar8w,hceiewypdcdnnu-uhfffaoysa,methyl 3,5-trimethoxy-2-nitrobenzoate PubChem CID: 78765 IUPAC Name: methyl 3,4,5-trimethoxy-2-nitrobenzoate SMILES: COC1=C(C(=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC)OC