missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Methoxy-5-(methylsulfonyl)benzoate 98.0+%, TCI America™

Catalog No. p-7132940
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19635G 5 g
M196325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19635G Supplier TCI America Supplier No. M19635G
Only null left

Chemical Identifiers

CAS 63484-12-8
Molecular Formula C10H12O5S
Molecular Weight (g/mol) 244.261
MDL Number MFCD01317545
InChI Key DSRPTFIZJVCPPS-UHFFFAOYSA-N
Synonym 2-Methoxy-5-(methylsulfonyl)benzoic Acid Methyl Ester
PubChem CID 6454622
IUPAC Name methyl 2-methoxy-5-methylsulfonylbenzoate
SMILES COC1=C(C=C(C=C1)S(=O)(=O)C)C(=O)OC

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 5 g
Formula Weight 244.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Methoxy-5-(methylsulfonyl)benzoate