accessibility menu, dialog, popup

UserName

CAS RN 63484-12-8

IUPAC Name: methyl 5-methanesulfonyl-2-methoxybenzoate
Synonyms: methyl 5-methanesulfonyl-2-methoxybenzoate
Molecular Weight (g/mol): 244.26
Molecular Formula: C10H12O5S
InChi Key: DSRPTFIZJVCPPS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1OC)S(C)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results