missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Isothiocyanatobenzoate 98.0+%, TCI America™

Catalog No. I057425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I057425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I057425G Supplier TCI America Supplier No. I057425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16024-82-1
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041062
InChI Key UNXVHBOJSCWVCD-UHFFFAOYSA-N
Synonym 2-isothiocyanato-benzoic acid methyl ester, 2-methoxycarbonylphenyl isothiocyanate, 2-methoxycarbonyl phenyl isothiocyanate, benzoic acid,2-isothiocyanato-, methyl ester, benzoic acid, 2-isothiocyanato-, methyl ester, acmc-209dkr, methyl2-isothiocyanatobenzoate, methyl 2-isothiocyanato benzoate, methyl 2-isothiocyanatobenzenecarboxylate, 2-isothiocyanatobenzoic acid methyl ester
PubChem CID 85243
IUPAC Name methyl 2-isothiocyanatobenzoate
SMILES COC(=O)C1=CC=CC=C1N=C=S

Specifications

Color Yellow
Quantity 25 g
Formula Weight 193.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl 2-Isothiocyanatobenzoate